6-(Trifluoromethoxy)benzo[d]thiazole 98%

Supplier: Ambeed
Warning

A176943-10G A176943-1G A176943-250MG A176943-25G A176943-5G
77096-666EA 133 USD
77096-666 77096-668 77096-670 77096-672 77096-674
6-(Trifluoromethoxy)benzo[d]thiazole 98%
6-(Trifluoromethoxy)benzo[d]thiazole

Precaución: for Research Use Only

Formula: C₈H₄F₃NOS
MDL Number: MFCD06659654
CAS Number: 876500-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR