2,8-Diazaspiro[4.5]decan-3-one hydrochloride 98%

Supplier: Ambeed
Warning

A176794-100MG A176794-250MG A176794-5G A176794-1G
77096-660EA 12 USD
77096-660 77096-662 77096-664 77593-122
2,8-Diazaspiro[4.5]decan-3-one hydrochloride 98%
2,8-Diazaspiro[4.5]decan-3-one hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₄N₂O·ClH
MDL Number: MFCD11520496
CAS Number: 945892-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR