2-Amino-3,5-dibromo-4-methylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dibromo-4-methylpyridin-2-amine

A176683-100G A176683-10G A176683-1G A176683-25G A176683-500G A176683-5G
77096-648EA 58.35 USD
77096-648 77096-650 77096-652 77096-654 77096-656 77096-658
2-Amino-3,5-dibromo-4-methylpyridine 97%
2-Amino-3,5-dibromo-4-methylpyridine

Precaución: for Research Use Only

Formula: C₅H₄Br₂N₂
MW: 265.9300 g/mol
MDL Number: MFCD01646065
CAS Number: 3430-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR