N-Acetylglycine(Ac-Gly-OH) 98%

Supplier: Ambeed
Warning

Synonyms: Ac-Gly-OH, N-Acetylglycine, Acetamidoacetic acid, Aceturic acid

A176529-1000G A176529-100G A176529-500G
77096-642EA 32 USD
77096-642 77096-644 77096-646
N-Acetylglycine(Ac-Gly-OH) 98%
N-Acetylglycine(Ac-Gly-OH)

Precaución: for Research Use Only

Formula: C₄H₇NO₃
MW: 117.1 g/mol
MDL Number: MFCD00004275
CAS Number: 543-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR