L-Aspartic aciddiethyl ester hydrochloride 95%

Supplier: Ambeed
Warning

A176340-100G A176340-10G A176340-25G A176340-500G A176340-5G
77096-628EA 61 USD
77096-628 77096-630 77096-632 77096-634 77096-636
L-Aspartic aciddiethyl ester hydrochloride 95%
L-Aspartic aciddiethyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₅NO₄·HCl
MDL Number: MFCD00038910
CAS Number: 16115-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR