Methyl 2-bromobenzo[d]thiazole-4-carboxylate 95%

Supplier: Ambeed
Warning

A175673-100MG A175673-1G A175673-250MG
77096-614EA 82.76 USD
77096-614 77096-616 77096-618
Methyl 2-bromobenzo[d]thiazole-4-carboxylate 95%
Methyl 2-bromobenzo[d]thiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂S
MW: 272.11 g/mol
MDL Number: MFCD22690014
CAS Number: 1440526-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR