1-Benzyl-3,4-dibromo-1H-pyrrole-2,5-dione 98+%

Supplier: Ambeed
Danger

A174714-100MG A174714-1G A174714-250MG A174714-5G
77096-598EA 15.92 USD
77096-598 77096-600 77096-602 77096-604
1-Benzyl-3,4-dibromo-1H-pyrrole-2,5-dione 98+%
1-Benzyl-3,4-dibromo-1H-pyrrole-2,5-dione

Precaución: for Research Use Only

Formula: C₁₁H₇Br₂NO₂
MW: 344.98 g/mol
MDL Number: MFCD03427193
CAS Number: 91026-00-5
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR