1-tert-Butyl 2-ethyl 5-oxopyrrolidine-1,2-dicarboxylate 96%

Supplier: Ambeed
Warning

A174582-1G A174582-250MG A174582-5G
77096-586EA 25.47 USD
77096-586 77096-588 77096-590
1-tert-Butyl 2-ethyl 5-oxopyrrolidine-1,2-dicarboxylate 96%
1-tert-Butyl 2-ethyl 5-oxopyrrolidine-1,2-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₅
MW: 257.28 g/mol
MDL Number: MFCD04113945
CAS Number: 251924-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR