(2-Amino-4-chloropyridin-3-yl)(phenyl)methanone 97%

Supplier: Ambeed
Danger

A174093-1G A174093-250MG
77096-574EA 301.3 USD
77096-574 77096-576
(2-Amino-4-chloropyridin-3-yl)(phenyl)methanone 97%
(2-Amino-4-chloropyridin-3-yl)(phenyl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₉ClN₂O
MW: 232.66 g/mol
MDL Number: MFCD17926323
CAS Number: 1203510-05-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR