1-(4-Bromophenyl)cyclopropanecarboxamide 95%

Supplier: Ambeed
Warning

A171808-100MG A171808-1G A171808-250MG
77096-536EA 149.59 USD
77096-536 77096-538 77096-540
1-(4-Bromophenyl)cyclopropanecarboxamide 95%
1-(4-Bromophenyl)cyclopropanecarboxamide

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrNO
MW: 240.10 g/mol
MDL Number: MFCD11644009
CAS Number: 847361-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR