1-(5-Bromo-6-fluoro-1H-indazol-1-yl)ethanone 97%

Supplier: Ambeed
Warning

A171063-100MG A171063-1G A171063-250MG A171063-50MG A171063-5G
77096-512EA 74.27 USD
77096-512 77096-514 77096-516 77096-518 77096-520
1-(5-Bromo-6-fluoro-1H-indazol-1-yl)ethanone 97%
1-(5-Bromo-6-fluoro-1H-indazol-1-yl)ethanone

Precaución: for Research Use Only

Formula: C₉H₆BrFN₂O
MW: 257.05 g/mol
MDL Number: MFCD17926217
CAS Number: 633335-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR