Methyl 3-(2-pyridyl)propiolate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-(2-Pyridyl)propiolate

A170707-100MG A170707-250MG
77096-498EA 51.99 USD
77096-498 77096-500
Methyl 3-(2-pyridyl)propiolate 95%
Methyl 3-(2-pyridyl)propiolate

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MW: 161.157 g/mol
MDL Number: MFCD02646628
CAS Number: 72764-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR