Boc-Abu-OH 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(tert-Butoxycarbonylamino)butyric acid, Boc-L-2-aminobutanoic acid, BOC-L-α-Aminobutyric acid, Boc-L-Abu-OH, (S)-2-(Boc-amino)butyric acid

A169982-1000G A169982-100G A169982-10G A169982-25G A169982-500G A169982-5G
77096-466EA 143 USD
77096-466 77096-468 77096-470 77096-472 77096-474 77096-476
Boc-Abu-OH 97%
Boc-Abu-OH

Precaución: for Research Use Only

Formula: C₉H₁₇NO₄
MDL Number: MFCD00037267
CAS Number: 34306-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR