(S)-(+)-Ketopinic acid 98%

Supplier: Ambeed
Warning

A169658-100MG A169658-1G A169658-5G
77096-458EA 6 USD
77096-458 77096-460 77567-488
(S)-(+)-Ketopinic acid 98%
(S)-(+)-Ketopinic acid

Precaución: For research use only.

Formula: C₁₀H₁₄O₃
MDL Number: MFCD00168053
CAS Number: 40724-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR