2-(1,2,4-Triazol-1-yl)aniline 95%

Supplier: Ambeed
Danger

A169171-1G A169171-250MG A169171-5G
77096-452EA 62.6 USD
77096-452 77096-454 77096-456
2-(1,2,4-Triazol-1-yl)aniline 95%
2-(1,2,4-Triazol-1-yl)aniline

Precaución: for Research Use Only

Formula: C₈H₈N₄
MW: 160.17 g/mol
MDL Number: MFCD08699291
CAS Number: 39876-84-1
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR