(2-Bromo-4-chlorophenyl)acetic acid 97%

Supplier: Ambeed
Warning

A168631-100G A168631-10G A168631-1G A168631-250MG A168631-25G A168631-5G
77096-440EA 367 USD
77096-440 77096-442 77096-444 77096-446 77096-448 77096-450
(2-Bromo-4-chlorophenyl)acetic acid 97%
(2-Bromo-4-chlorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
MDL Number: MFCD09032951
CAS Number: 52864-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR