4-Benzyloxy-2-chlorophenylboronic acid 97%

Supplier: Ambeed
Warning

A168251-1G A168251-250MG A168251-5G
77096-426EA 27.59 USD
77096-426 77096-428 77096-430
4-Benzyloxy-2-chlorophenylboronic acid 97%
4-Benzyloxy-2-chlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BClO₃
MW: 262.497 g/mol
MDL Number: MFCD06801734
CAS Number: 1315341-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR