H-Lys(crotonyl)-OH 95%

Supplier: Ambeed
Warning

Synonyms: (S,E)-2-Amino-6-(but-2-enamido)hexanoic acid

A168042-100MG A168042-250MG
77096-414EA 35.01 USD
77096-414 77096-416
H-Lys(crotonyl)-OH 95%
H-Lys(crotonyl)-OH

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₃
MW: 214.26 g/mol
MDL Number: MFCD22381072
CAS Number: 1338823-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR