7-Fluorobenzo[b]furan 97%

Supplier: Ambeed
Warning

A167722-100MG A167722-250MG A167722-5G
77096-408EA 15 USD
77096-408 77096-410 77096-412
7-Fluorobenzo[b]furan 97%
7-Fluorobenzo[b]furan

Precaución: for Research Use Only

Formula: C₈H₅FO
MDL Number: MFCD15143817
CAS Number: 24410-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR