C-(7-Methyl-[1,2,4]triazolo[4,3-a]pyridin-3-yl)-methylamine 95%

Supplier: Ambeed
Danger

A167349-1G A167349-250MG
77096-394EA 145 USD
77096-394 77096-396
C-(7-Methyl-[1,2,4]triazolo[4,3-a]pyridin-3-yl)-methylamine 95%
C-(7-Methyl-[1,2,4]triazolo[4,3-a]pyridin-3-yl)-methylamine

Precaución: for Research Use Only

Formula: C₈H₁₀N₄
MDL Number: MFCD09994896
CAS Number: 1020033-70-8
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR