(S)-Isopropyl 2-(((S)-((((R)-1-(6-amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)(phenoxy)phosphoryl)amino)propanoate fumarate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tenofovir alafenamide fumarate
A167277-10MG
A167277-1MG
A167277-250MG
A167277-25MG
A167277-50MG
A167277-5G
A167277-5MG
A167277-1G
77096-380EA
18
USD
77096-380
77096-382
77096-384
77096-386
77096-388
77096-390
77096-392
77685-360
(S)-Isopropyl 2-(((S)-((((R)-1-(6-amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)(phenoxy)phosphoryl)amino)propanoate fumarate 98%
(S)-Isopropyl 2-(((S)-((((R)-1-(6-amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)(phenoxy)phosphoryl)amino)propanoate fumarate
Precaución: for Research Use Only
Formula:
C₂₅H₃₃N₆O₉P |
MDL Number:
MFCD28167823 CAS Number: 379270-38-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...