4-Bromo-2-chlorobenzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-bromobenzaldehyde

A167193-100G A167193-10G A167193-1G A167193-25G A167193-500G A167193-5G
77096-368EA 82.76 USD
77096-368 77096-370 77096-372 77096-374 77096-376 77096-378
4-Bromo-2-chlorobenzaldehyde 98%
4-Bromo-2-chlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrClO
MW: 219.46 g/mol
MDL Number: MFCD08741413
CAS Number: 158435-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR