3,6-Di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione 95%

Supplier: Ambeed
Warning

Synonyms: 3,6-Di(thiophen-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione

A166415-1G A166415-250MG A166415-25G A166415-5G
77096-334EA 39.26 USD
77096-334 77096-336 77096-338 77096-340
3,6-Di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione 95%
3,6-Di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione

Precaución: for Research Use Only

Formula: C₁₄H₈N₂O₂S₂
MW: 300.35 g/mol
MDL Number: MFCD20257907
CAS Number: 850583-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR