Propargyl amine 98%

Supplier: Ambeed
Danger

Synonyms: Monopropagyl amine, Prop-2-ynylamine, 3-Aminopropyne, Prop-2-yn-1-amine

A166127-10G A166127-1G A166127-25G A166127-5G
77096-314EA 15 USD
77096-314 77096-316 77096-318 77096-320
Propargyl amine 98%
Propargyl amine

Precaución: for Research Use Only

Formula: C₃H₅N
MW: 55.08 g/mol
Boiling Pt: 83
MDL Number: MFCD00008198
CAS Number: 2450-71-7
UN: 3286
ADR: 3,6.1,8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR