2-Hydroxy-3-(5-(morpholinomethyl)pyridin-2-yl)-1H-indole-5-carbonitrile 98+%

Supplier: Ambeed
Warning

Synonyms: AZD1080

A166126-10MG A166126-2MG A166126-5MG A166126-1MG A166126-50MG
77096-308EA 33.95 USD
77096-308 77096-310 77096-312 77611-618 77685-352
2-Hydroxy-3-(5-(morpholinomethyl)pyridin-2-yl)-1H-indole-5-carbonitrile 98+%
2-Hydroxy-3-(5-(morpholinomethyl)pyridin-2-yl)-1H-indole-5-carbonitrile

Precaución: for Research Use Only

Formula: C₁₉H₁₈N₄O₂
MW: 334.37 g/mol
MDL Number: MFCD12031593
CAS Number: 612487-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR