SGC707 98+%

Supplier: Ambeed
Warning

Synonyms: 1-(Isoquinolin-6-yl)-3-(2-oxo-2-(pyrrolidin-1-yl)ethyl)urea

A165657-10MG A165657-1MG A165657-50MG A165657-5MG
77096-294EA 35.01 USD
77096-294 77096-296 77096-298 77096-300
SGC707 98+%
SGC707

Precaución: for Research Use Only

Formula: C₁₆H₁₈N₄O₂
MW: 298.34 g/mol
MDL Number: MFCD28411624
CAS Number: 1687736-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR