Methyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropionate 98%

Supplier: Ambeed
Warning

A165357-100MG A165357-1G A165357-1MG A165357-250MG A165357-5G
77096-272EA 7 USD
77096-272 77096-274 77096-276 77096-278 77096-280
Methyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropionate 98%
Methyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropionate

Precaución: for Research Use Only

Formula: C₁₇H₁₇NO₄
MDL Number: MFCD00673331
CAS Number: 32981-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR