4-Fluoro-3-nitrophenol 98%

Supplier: Ambeed
Danger

A165013-100G A165013-10G A165013-1G A165013-5G A165013-25G A165013-250MG
77096-264EA 379 USD
77096-264 77096-266 77096-268 77096-270 77685-342 77901-839
4-Fluoro-3-nitrophenol 98%
4-Fluoro-3-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄FNO₃
MDL Number: MFCD04038714
CAS Number: 2105-96-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR