Pyrimido[4,5-d]pyridazine-2,4(1H,3H)-dione 97%

Supplier: Ambeed
Warning

A163118-100MG A163118-250MG
77096-210EA 175 USD
77096-210 77096-212
Pyrimido[4,5-d]pyridazine-2,4(1H,3H)-dione 97%
Pyrimido[4,5-d]pyridazine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C6H4N4O2
MDL Number: MFCD11111471
CAS Number: 17257-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR