Oxazolo[4,5-b]pyridin-2(3H)-one 97%

Supplier: Ambeed
Warning

A163068-100G A163068-1G A163068-5G A163068-10G
77096-202EA 50 USD
77096-202 77096-204 77096-206 77567-378
Oxazolo[4,5-b]pyridin-2(3H)-one 97%
Oxazolo[4,5-b]pyridin-2(3H)-one

Precaución: For research use only.

Formula: C₆H₄N₂O₂
MDL Number: MFCD00204215
CAS Number: 60832-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR