(R)-b-Amino-4-bromo-benzeneethanol 95%

Supplier: Ambeed
Warning

Synonyms: (R)-2-Amino-2-(4-bromophenyl)ethanol

A162611-250MG A162611-5G
77096-200EA 99.73 USD
77096-200 77685-332
(R)-b-Amino-4-bromo-benzeneethanol 95%
(R)-b-Amino-4-bromo-benzeneethanol

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
MW: 216.075 g/mol
MDL Number: MFCD09256373
CAS Number: 354153-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR