2-Amino-3-bromo-4-picoline 97%

Supplier: Ambeed
Warning

A162409-10G A162409-1G A162409-250MG A162409-5G
77096-192EA 146 USD
77096-192 77096-194 77096-196 77096-198
2-Amino-3-bromo-4-picoline 97%
2-Amino-3-bromo-4-picoline

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MDL Number: MFCD08061297
CAS Number: 40073-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR