6-Bromo-7-chloro-1H-benzo[d]imidazole 97%

Supplier: Ambeed
Warning

A161266-1G A161266-250MG A161266-5G
77096-168EA 116 USD
77096-168 77096-170 77096-172
6-Bromo-7-chloro-1H-benzo[d]imidazole 97%
6-Bromo-7-chloro-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.47 g/mol
MDL Number: MFCD12828061
CAS Number: 1008361-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR