4-Fluoro-3-(trifluoromethyl)phenylacetonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 4-fluoro-3-(trifluoromethyl)phenylacetonitrile

A160709-250MG A160709-5G
77096-142EA 6.37 USD
77096-142 77096-144
4-Fluoro-3-(trifluoromethyl)phenylacetonitrile 98%
4-Fluoro-3-(trifluoromethyl)phenylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₅F₄N
MW: 203.14 g/mol
MDL Number: MFCD00061183
CAS Number: 220239-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR