Dibenzyl azodicarboxylate 96%

Supplier: Ambeed
Danger

A160551-1G A160551-5G A160551-25G
77096-136EA 53.05 USD
77096-136 77096-138 77096-140 77567-338
Dibenzyl azodicarboxylate 96%
Dibenzyl azodicarboxylate

Precaución: For research use only.

Formula: C₁₆H₁₄N₂O₄
MDL Number: MFCD00016737
CAS Number: 2449-05-0
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR