PHT-427 98%

Supplier: Ambeed
Warning

Synonyms: 4-Dodecyl-N-(1,3,4-thiadiazol-2-yl)benzenesulphonamide

A160471-10MG A160471-1MG
77096-132EA 40 USD
77096-132 77096-134
PHT-427 98%
PHT-427

Precaución: for Research Use Only

Formula: C₂₀H₃₁N₃O₂S₂
MW: 409.61 g/mol
MDL Number: MFCD18384970
CAS Number: 1191951-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR