3-Bromo-6-iodo-o-xylene 95%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2,3-dimethyl-4-iodobenzene

A160008-1G A160008-25G A160008-5G
77096-116EA 21.22 USD
77096-116 77096-118 77096-120
3-Bromo-6-iodo-o-xylene 95%
3-Bromo-6-iodo-o-xylene

Precaución: for Research Use Only

Formula: C₈H₈BrI
MW: 310.96 g/mol
MDL Number: MFCD07778967
CAS Number: 1160573-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR