1-Methyl-1H-indole-5-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-5-indolecarboxylic Acid

A159539-5G A159539-1G
77096-108EA 178.24 USD
77096-108 77685-306
1-Methyl-1H-indole-5-carboxylic acid 98%
1-Methyl-1H-indole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
MDL Number: MFCD03839859
CAS Number: 186129-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR