2-[4-(Trifluoromethyl)phenyl]pyridine 98%

Supplier: Ambeed
Warning

A157722-1G A157722-250MG
77096-068EA 32 USD
77096-068 77096-070
2-[4-(Trifluoromethyl)phenyl]pyridine 98%
2-[4-(Trifluoromethyl)phenyl]pyridine

Precaución: for Research Use Only

Formula: C₁₂H₈F₃N
MDL Number: MFCD08235168
CAS Number: 203065-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR