Potassium oxonate 98%

Supplier: Ambeed
Warning

Synonyms: Potassium Allantoxanate, Oxonic acid Potassium Salt, Potassium 4,6-Dihydroxy-1,3,5-triazine-2-carboxylate, Allantoxanic acid Potassium Salt, Potassium 4,6-dioxo-1,4,5,6-tetrahydro-1,3,5-triazine-2-carboxylate

A157215-100G A157215-10G A157215-1G A157215-250MG A157215-25G A157215-5G
77096-026EA 112.46 USD
77096-026 77096-028 77096-030 77096-032 77096-034 77096-036
Potassium oxonate 98%
Potassium oxonate

Precaución: for Research Use Only

Formula: C₄H₃N₃O₄
MW: 195.18 g/mol
MDL Number: MFCD00010565
CAS Number: 2207-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR