2,4,6-Tris(3-bromophenyl)-1,3,5-triazine 95%

Supplier: Ambeed
Warning

A157015-100G A157015-1G A157015-25G A157015-5G
77096-018EA 615.33 USD
77096-018 77096-020 77096-022 77096-024
2,4,6-Tris(3-bromophenyl)-1,3,5-triazine 95%
2,4,6-Tris(3-bromophenyl)-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₂₁H₁₂Br₃N₃
MW: 546.06 g/mol
MDL Number: MFCD28964709
CAS Number: 890148-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR