(3,4,5-Trifluorophenyl)acetonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 2-(3,4,5-Trifluorophenyl)acetonitrile, 3,4,5-Trifluorobenzyl cyanide

A156647-5G A156647-1G
77096-008EA 55.17 USD
77096-008 77685-291
(3,4,5-Trifluorophenyl)acetonitrile 97%
(3,4,5-Trifluorophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₄F₃N
MW: 171.12 g/mol
MDL Number: MFCD00083531
CAS Number: 220228-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR