Dicyclohexyl-3,4,3',4'-tetracarboxylic Dianhydride 97%

Supplier: Ambeed
Warning

A156120-100G A156120-10G A156120-25G A156120-5G
77095-992EA 158 USD
77095-992 77095-994 77095-996 77095-998
Dicyclohexyl-3,4,3',4'-tetracarboxylic Dianhydride 97%
Dicyclohexyl-3,4,3',4'-tetracarboxylic Dianhydride

Precaución: for Research Use Only

Formula: C₁₆H₁₈O₆
MDL Number: MFCD23134595
CAS Number: 122640-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR