4-Bromo-1,2-dichlorobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3,4-dichlorobenzene

A155814-1000G A155814-5G A155814-25G A155814-100G A155814-500G
77095-968EA 199.45 USD
77095-968 77095-970 77300-148 77611-482 77611-484
4-Bromo-1,2-dichlorobenzene 98%
4-Bromo-1,2-dichlorobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
MDL Number: MFCD00040849
CAS Number: 18282-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR