(2-Amino-9H-fluoren-9-yl)methanol 95%

Supplier: Ambeed
Warning

A155688-100MG A155688-1G A155688-250MG A155688-5G
77095-954EA 55.17 USD
77095-954 77095-956 77095-958 77095-960
(2-Amino-9H-fluoren-9-yl)methanol 95%
(2-Amino-9H-fluoren-9-yl)methanol

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO
MW: 211.26 g/mol
MDL Number: MFCD24646252
CAS Number: 162021-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR