2-(Trifluoromethyl)furan-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A155479-100MG A155479-250MG
77095-950EA 25.47 USD
77095-950 77095-952
2-(Trifluoromethyl)furan-3-carboxylic acid 95%
2-(Trifluoromethyl)furan-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃F₃O₃
MW: 180.081 g/mol
MDL Number: MFCD03662621
CAS Number: 536718-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR