5-Chloro-2,4-dihydroxybenzaldehyde 95%

Supplier: Ambeed
Warning

A155186-100MG A155186-1G A155186-250MG A155186-25G A155186-5G
77095-924EA 16.98 USD
77095-924 77095-926 77095-928 77095-930 77095-932
5-Chloro-2,4-dihydroxybenzaldehyde 95%
5-Chloro-2,4-dihydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅ClO₃
MW: 172.56 g/mol
MDL Number: MFCD18324145
CAS Number: 131088-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR