2-Fluoro-5-((4-oxo-3,4-dihydrophthalazin-1-yl)methyl)benzonitrile 97%

Supplier: Ambeed
Warning

A154503-1G A154503-250MG A154503-25G A154503-5G
77095-910EA 25.47 USD
77095-910 77095-912 77095-914 77095-916
2-Fluoro-5-((4-oxo-3,4-dihydrophthalazin-1-yl)methyl)benzonitrile 97%
2-Fluoro-5-((4-oxo-3,4-dihydrophthalazin-1-yl)methyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₆H₁₀FN₃O
MW: 279.269 g/mol
MDL Number: MFCD28133450
CAS Number: 1021298-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR