3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile 97%

Supplier: Ambeed
Warning

A153961-100MG A153961-1G A153961-250MG
77095-904EA 37.14 USD
77095-904 77095-906 77095-908
3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile 97%
3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile

Precaución: for Research Use Only

Formula: C₁₂H₁₄BClN₂O₂
MW: 264.52 g/mol
MDL Number: MFCD16995199
CAS Number: 1220219-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR