tert-Butyl 5-bromovalerate 95%

Supplier: Ambeed
Warning

A153912-10G A153912-1G A153912-250MG A153912-25G A153912-5G A153912-100G
77095-896EA 39 USD
77095-896 77095-898 77095-900 77095-902 77549-954 77685-275
tert-Butyl 5-bromovalerate 95%
tert-Butyl 5-bromovalerate

Precaución: for Research Use Only

Formula: C₉H₁₇BrO₂
MW: 237.13 g/mol
MDL Number: MFCD26398445
CAS Number: 88987-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR